Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polypeptide GalNac Transferase 10/GALNT10 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177058G
Description
Polypeptide GalNac Transferase 10/GALNT10 Polyclonal antibody specifically detects Polypeptide GalNac Transferase 10/GALNT10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Polypeptide GalNac Transferase 10/GALNT10 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
55568 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
DKFZp586H0623, EC 2.4.1.41, FLJ00205, FLJ11715, GalNAc transferase 10, GalNAc-T10, GALNACT10, Polypeptide GalNAc transferase 10, polypeptide N-acetylgalactosaminyltransferase 10, PPGALNACT10, pp-GaNTase 10, PPGANTASE10, Protein-UDP acetylgalactosaminyltransferase 10, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 10, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 10 (GalNAc-T10) | |
Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human GALNT10. The immunogen is located within amino acids 530 - 580 of GALNT10. Amino Acid Squence: KGNQLWKYRKDKTLYHPV | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Polypeptide GalNac Transferase 10/GALNT10 Antibody, DyLight 488, Novus Biologicals™ > 0.1 mL; DyLight 488
Spot an opportunity for improvement?Share a Content Correction