Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRRS Virus GP5 Protein Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111970

Description
Porcine reproductive and respiratory syndrome virus (PRRSV) GP5 glycosylated envelope protein.
Specifications
PRRS Virus GP5 Protein | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Western Blot | |
0.57 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Recombinant protein encompassing a sequence within the center region of PRRS virus GP5 protein. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PRRS Virus GP5 Protein Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction