Learn More
PTPN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA542722
Description
Peptide sequence: ESGSLNPDHG PAVIHCSAGI GRSGTFSLVD TCLVLMEKGD DINIKQVLLN Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
PTPN2 Polyclonal antibody specifically detects PTPN2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.
Specifications
PTPN2 | |
Polyclonal | |
Unconjugated | |
PTPN2 | |
AI325124; hypothetical protein LOC540888; MPTP; protein tyrosine phosphatase non-receptor type 2; protein tyrosine phosphatase, non-receptor type 2; protein-tyrosine phosphatase PTP-2; Protein-tyrosine phosphatase PTP-S; PTN2; PTPN 2; PTPN2; Ptpt; T cell protein-tyrosine phosphatase; TC PTP; T-cell protein tyrosine phosphatase; T-cell protein-tyrosine phosphatase; TCELLPTP; TCPTP; TC-PTP; tyrosine-phosphatase; Tyrosine-protein phosphatase non-receptor type 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5771 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P17706 | |
PTPN2 | |
Synthetic peptide directed towards the middle region of human PTPN2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.