Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Quidel Sofia™ RSV FIA Immunofluorescence Kit Catalog No.: 23043081

Quidel Sofia™ RSV FIA Immunofluorescence Kit
SDP

Catalog No. 23043081
3 x 25 Tests
Click to view available options
Quantity:
1 x 25 Tests
3 x 25 Tests
This item is not returnable. View return policy

Catalog No. 23-043-081

Supplier: Quidel 20264

This item is not returnable. View return policy

Includes

RSV FIA 25-test Kit Contents:

  • Individually Packaged Cassettes (25): Mouse monoclonal anti-RSV antibodies
  • Reagent Tubes (25): Lyophilized buffer with detergents and reducing agents
  • Reagent Solution (25): Vials with salt solution
  • Sterile Nasopharyngeal Swabs (25)
  • Large, Pink Fixed Volume Pipettes (25)
  • Small, Clear Fixed Volume Pipettes (25)
  • RSV Positive Control Swab (1): Swab is coated with non-infectious RSV antigen
  • Negative Control Swab (1): Swab is coated with heat-inactivated, non-infectious Streptococcus C antigen
  • Package Insert (1)
  • Quick Reference Instructions (1)
  • QC Card (located on kit box)
  • Printer Paper (1)

For rapid qualitative detection of respiratory syncytial virus (RSV) nucleoprotein antigens

 

  • Use with Sofia Analyzer only
  • Test nasopharyngeal swab and nasopharyngeal aspirate/wash specimens taken directly from symptomatic patients less than 19 years of age
  • When run in conjunction with Sofia Influenza A+B FIA, a single specimen may be used for both tests
  • Accurate results within 15 mins.
  • 25 tests per kit

 

Applications:   IVD

CLIA Complexity: This test is CLIA moderately complex for pediatric patients 7 to less than 19 years of age. It is CLIA waived for children less than 7 years of age.

Order Info

Order eligibility is based upon executed contractual agreement. Please reach out to your Fisher HealthCare Account Representative for more information.

Certifications

CE Marked; CLIA Complexity: Waived

Specifications

Clia Complexity Waived
Detectable Analytes Respiratory Syncytial Virus (RSV) Antigen
Sample Type Nasopharyngeal swab and nasopharyngeal aspirate/wash specimens
Type Sofia™ RSV FIA Immunofluorescence Test Kit
For Use With (Application) IVD
Includes Individually packaged cassettes 25, reagent tubes 25, reagent solution 25, sterile nasopharyngeal swabs 25, large, pink fixed volume pipettes 25, small, clear fixed volume pipettes 25, RSV positive control swab 1, negative control swab 1, package insert 1, quick reference instructions 1, QC card, printer Paper 1
Quantity 3 x 25 Tests
Test Time >15 min.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Quidel Sofia™ RSV FIA Immunofluorescence Kit >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.