Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rad51 Rabbit anti-Human, Mouse, Rat, Clone: JM54-26, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275640
Description
Rad51 Monoclonal specifically detects Rad51 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Rad51 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:1000 | |
BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A | |
Rabbit | |
37 kDa | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
JM54-26 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
RAD51 | |
Synthetic peptide within Human Rad51 aa 1-50/ 339. (SwissProt: Q06609 Human; SwissProt: Q08297 Mouse; SwissProt: B5DF04 Rat) | |
Protein A purified | |
Cancer, DNA Repair, Homologous Recombination, modENCODE Antibodies | |
5888.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Rad51 Rabbit anti-Human, Mouse, Rat, Clone: JM54-26, Novus Biologicals™ > 100 μl; Unconjugated
Spot an opportunity for improvement?Share a Content Correction