Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Follistatin (aa 30-344) Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose and with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Description
The Recombinant Human Follistatin (aa 30-344) Protein is derived from CHO. The Recombinant Human Follistatin (aa 30-344) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS with Trehalose and with BSA as a carrier protein. |
Gene ID (Entrez) | 10468 |
Molecular Weight (g/mol) | 34.7 kDa |
Name | Follistatin 315 aa 30-344 |
Quantity | 25 μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human Follistatin protein Gly30-Trp344 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Human Follistatin (aa 30-344) Protein >
Spot an opportunity for improvement?Share a Content Correction