Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGL2 Antibody (OTI10F2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273875F
Description
RGL2 Monoclonal specifically detects RGL2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
RGL2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
RGL2 | |
Recombinant protein fragment corresponding to amino acids 547-777 of human RGL2 (NP_004752) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Immunohistochemistry | |
OTI10F2 | |
Immunohistochemistry | |
GDS-related protein, HKE1.5, KE1.5, RAB2, member RAS oncogene family-like, RAB2LRas-associated protein RAB2L, ral guanine nucleotide dissociation stimulator-like 2, RalGDS-like 2, RalGDS-like factor | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
5863.0 | |
Store at 4°C in the dark. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RGL2 Antibody (OTI10F2), FITC, Novus Biologicals™ > 0.1 mL; FITC
Spot an opportunity for improvement?Share a Content Correction