Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SGK3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5117289

Description
Centrifuge briefly prior to opening.
SGKL (SGK3) is a serine/threonine protein kinase which is similar to serum- and glucocorticoid-induced protein kinase (SGK). Unlike SGK, SGKL is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. The protein phosphorylates several target proteins and has a role in neutral amino acid transport and activation of potassium and chloride channels. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
SGK3 | |
Polyclonal | |
Unconjugated | |
SGK3 | |
2510015P22Rik; A330005P07Rik; CISK; cytokine-independent survival kinase; DKFZp781N0293; fy; fz; Serine/threonine-protein kinase Sgk3; serum glucocorticoid regulated kinase 3; serum/glucocorticoid regulated kinase 3; serum/glucocorticoid regulated kinase family member 3; serum/glucocorticoid regulated kinase family, member 3; serum/glucocorticoid-regulated kinase 3; Serum/glucocorticoid-regulated kinase-like; SGK2; SGK3; Sgkl | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23678 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96BR1 | |
SGK3 | |
Recombinant protein encompassing a sequence within the N-terminus region of human SGK3. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SGK3 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction