Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Neutralite Avidin
SDP

Catalog No. OB720009
Click to view available options
Quantity:
0.1 mg
0.5 mg
1 mg
5 mg
1 mL
Conjugate:
AP
APC
Cy3
Cy5
FITC (Fluorescein)
HRP
R-phycoerythrin
Rhodamine
Texas Red
Unconjugated
beta-Galactosidase
pH:
7.4
8
8.1
Color:
Blue
Blue to Violet
Clear
Colorless to Light Brown
Light to Dark Amber
Light to Dark Pink
Light to Dark Yellow
Red to Dark Violet
Reagents contain sodium azide. Sodium azide is very toxic if ingested or inhaled. Avoid contact with skin, eyes, or clothing. Wear eye or face protection when handling. If skin or eye contact occurs, wash with copious amounts of water. If ingested or inhaled, contact a physician immediately. Sodium azide yields toxic hydrazoic acid under acidic conditions. Dilute azide-containing compounds in running water before discarding to avoid accumulation of potentially explosive deposits in lead or copper plumbing.
Reagents contain sodium azide. Sodium azide is very toxic if ingested or inhaled. Avoid contact with skin, eyes, or clothing. Wear eye or face protection when handling. If skin or eye contact occurs, wash with copious amounts of water. If ingested or inhaled, contact a physician immediately. Sodium azide yields toxic hydrazoic acid under acidic conditions. Dilute azide-containing compounds in running water before discarding to avoid accumulation of potentially explosive deposits in lead or copper plumbing.

Neutralized, deglycosylated avidin purified from egg white

For Research Use Only. Not for Diagnostic or Therapeutic Use.

Flow cytometry; Enzyme-Linked-Immunosorbent-Assay (ELISA); Immunoblotting; Immunohistochemistry

Order Info

To ensure lot-to-lot consistency, each batch of product is tested by flow cytometry, particle concentration, FLISA or ELISA to conform to characteristics of a standard reference reagent.

Specifications

Color Light to Dark Pink
Concentration 0.5 mg/mL
Conjugate R-phycoerythrin
pH 7.4
Quantity 0.5 mg
Source Egg White
Content And Storage 2°C to 8°C
Form Liquid
For Use With (Application) ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry
Specificity d-Biotin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.