Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SouthernBiotech™ Neutralite Avidin

Click to view available options
Quantity:
0.1 mg
0.5 mg
1 mg
5 mg
1 mL
Conjugate:
AP
APC
Cy3
Cy5
FITC (Fluorescein)
HRP
R-phycoerythrin
Rhodamine
Texas Red
Unconjugated
beta-Galactosidase
pH:
7.4
8
8.1
Color:
Blue
Blue to Violet
Clear
Colorless to Light Brown
Light to Dark Amber
Light to Dark Pink
Light to Dark Yellow
Red to Dark Violet
Description
For Research Use Only. Not for Diagnostic or Therapeutic Use.
Flow cytometry; Enzyme-Linked-Immunosorbent-Assay (ELISA); Immunoblotting; Immunohistochemistry
Order Info
To ensure lot-to-lot consistency, each batch of product is tested by flow cytometry, particle concentration, FLISA or ELISA to conform to characteristics of a standard reference reagent.
Specifications
Specifications
Color | Light to Dark Yellow |
Concentration | 0.5 mg/mL |
Conjugate | Cy3 |
pH | 7.4 |
Quantity | 0.1 mg |
Source | Egg White |
Content And Storage | 2°C to 8°C |
Form | Liquid |
For Use With (Application) | ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry |
Specificity | d-Biotin |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SouthernBiotech™ Neutralite Avidin >
Spot an opportunity for improvement?Share a Content Correction