Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPATA16 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA112696
Description
This gene encodes a testis-specific protein that belongs to the tetratricopeptide repeat-like superfamily. The encoded protein localizes to the Golgi apparatus and may play a role in spermatogenesis.
Specifications
SPATA16 | |
Polyclonal | |
Unconjugated | |
SPATA16 | |
4921511F01Rik; 4930503K02Rik; CDC68; FACT; FACTP140; FLJ10857; FLJ14010; NYD-SP12; RGD1563726; Spata16; spermatogenesis associated 16; spermatogenesis-associated protein 16; spermatogenesis-related protein; SPGF6; SUPT16H; testicular tissue protein Li 185; testis development protein NYD-SP12; testis-specific Golgi protein | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
70862, 83893 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ChIP Assay, Western Blot | |
1 mg/mL | |
PBS with 0.08% sodium azide | |
Q8C636, Q9BXB7 | |
SPATA16 | |
Recombinant protein consisting of amino acids 1-329 of the human SPT16 protein conjugated to KLH. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SPATA16 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction