Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STEAP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5102321

Description
Antibody detects endogenous levels of total STEA3.
The six-transmembrane epithelial antigen of prostate 3 (STEAP3) is a member of a family of metalloreductases identified as cell-surface antigens in prostate tissue. Similar to two other members of the STEAP family (STEAP 2 and STEAP4), STEAP3 promotes both iron and copper reduction. STEAP3 is highly expressed in hematopoietic tissues and colocalizes with the transferrin endosome. Overexpression of STEAP3 stimulates iron reduction; mice lacking STEAP3 are deficient in erythroid ferrireductase activity, suggesting that STEAP3 is an endosomal ferrireductase required for transferrin-dependent iron uptake in erythroid cells.
Specifications
STEAP3 | |
Polyclonal | |
Unconjugated | |
STEAP3 | |
1010001D01Rik; AHMIO2; dudlin 2; dudlin-2; dudulin 2; dudulin-2; hpHyde; hTSAP6; Metalloreductase STEAP3; pHyde; Protein nm1054; six-transmembrane epithelial antigen of prostate 3; STEAP family member 3; STEAP family member 3, metalloreductase; STEAP3; STEAP3 metalloreductase; STMP3; TSAP6; tumor suppressor pHyde; tumor suppressor-activated pathway protein 6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
170824, 55240, 68428 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5RKL5, Q658P3, Q8CI59 | |
STEAP3 | |
A synthesized peptide derived from human STEAP3(Accession Q658P3), corresponding to amino acid residues D16-R66. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
STEAP3 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction