Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Talin1 Antibody (93E12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Talin1 |
---|---|
Clone | 93E12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Description
Talin1 Monoclonal antibody specifically detects Talin1 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
Talin1 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Cell Biology, Cellular Markers | |
PBS | |
7094 | |
IgG1 | |
Protein A purified |
93E12 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
ILWEQ, KIAA1027, talin 1, talin-1, TLN | |
Recombinant protein fragment | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Talin1 Antibody (93E12) - BSA Free, Novus Biologicals™