Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Human p53, Recombinant Protein, Invitrogen™ Catalog No.: PIRP39232

Human p53, Recombinant Protein, Invitrogen™

Catalog No. PIRP39232
10 μg
Click to view available options
Quantity:
10 μg

Catalog No. PIRP39232

Supplier: Thermo Scientific RP39232

Recombinant Protein

p53 wild Type Recombinant Protein, RP-39232, from Invitrogen.

This gene encodes a tumor suppressor protein containing transcriptional activation, DNA binding, and oligomerization domains. The encoded protein responds to diverse cellular stresses to regulate expression of target genes, thereby inducing cell cycle arrest, apoptosis, senescence, DNA repair, or changes in metabolism. Mutations in this gene are associated with a variety of human cancers, including hereditary cancers such as Li-Fraumeni syndrome. Alternative splicing of this gene and the use of alternate promoters result in multiple transcript variants and isoforms. Additional isoforms have also been shown to result from the use of alternate translation initiation codons This product is a recombinant protein corresponding to full-length human p53 wild type. On SDS-PAGE commassie blue stained gel, the purified recombinant protein shows a band at 81kDa.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04637
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 0.05M tris HCl with 0.01M L-glutathione and no preservative; pH 7.5
Gene ID (Entrez) 7157
Name Human p53
pH Range 7.5
Quantity 10 μg
Source E. coli
Storage Requirements -20°C or -80°C if preferred
Regulatory Status RUO
Gene Alias antigen NY-CO-13, cellular tumor antigen p53, p53 tumor suppressor, phosphoprotein p53, transformation-related protein 53, TP53
Common Name Human p53
Gene Symbol TP53
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitro research only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Human p53, Recombinant Protein, Invitrogen™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.