Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM66 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA531588
Description
Recommended positive controls: 293T, Jurkat, Raji. Predicted reactivity: Mouse (85%), Rat (84%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Saraf encodes a protein that is a negative regulator of store-operated Ca(2+) entry (SOCE) involved in protecting cells from Ca(2+) overfilling. In response to cytosolic Ca(2+) elevation after endoplasmic reticulum Ca(2+) refilling, promotes a slow inactivation of STIM (STIM1 or STIM2)-dependent SOCE activity: possibly act by facilitating the de-oligomerization of STIM to efficiently turn off ORAI when the endoplasmic reticulum lumen is filled with the appropriate Ca(2+) levels, and thus preventing the overload of the cell with excessive Ca(2+) ions.
Specifications
TMEM66 | |
Polyclonal | |
Unconjugated | |
SARAF | |
1810045K07Rik; arrestin-E; FOAP-7; HBV XAg-transactivated protein 3; HBV X-transactivated gene 3 protein; HSPC035; NPD003; Protein FOAP-7; PSEC0019; Saraf; SARAF long isoform; SARAF short isoform; SOCE-associated regulatory factor; store-operated calcium entry associated regulatory factor; store-operated calcium entry-associated regulatory factor; testicular secretory protein Li 59; TMEM66; Transmembrane protein 66; UNQ1967/PRO4499; XTP3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51669 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96BY9 | |
SARAF | |
Recombinant fragment corresponding to a region within amino acids 1 and 211 of Human Transmembrane protein 66 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TMEM66 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction