Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEPAI Antibody (PMEPA1/2697), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279892V
Description
TMEPAI Monoclonal specifically detects TMEPAI in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TMEPAI | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PMEPA1 | |
Recombinant full-length human TMEPAI protein (Uniprot: Q96W9) | |
0.1 mL | |
Signal Transduction | |
Human | |
IgG κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
PMEPA1/2697 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
prostate transmembrane protein, androgen induced 1, Solid tumor-associated 1 protein, STAG1prostate androgen induced RNA, transmembrane prostate androgen-induced protein | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TMEPAI Antibody (PMEPA1/2697), DyLight 405, Novus Biologicals™ > 0.1 mL; DyLight 405
Spot an opportunity for improvement?Share a Content Correction