Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troponin T Type 2 (cardiac) Antibody (BGN/1288/44), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066490AF532
Description
Troponin T Type 2 (cardiac) Monoclonal specifically detects Troponin T Type 2 (cardiac) in Human samples. It is validated for Western Blot, ELISA.Specifications
Troponin T Type 2 (cardiac) | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
TNNT2 | |
HMW conjugate | |
Primary | |
NB100-66490 recognises Troponin T Type 2 (cardiac) isoform 1. In Western blotting, this reacts with reduced purified cardiac isoform, but does not react with the skeletal isoform. | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
BGN/1288/44 | |
Western Blot, ELISA | |
cardiac, CMD1D, CMPD2, dilated 1D (autosomal dominant), hypertrophic 2, MGC3889, RCM3, TnTc, troponin T type 2 (cardiac), troponin T, cardiac muscle | |
Mouse | |
0.1 mL | |
7139 | |
Human | |
IgG1 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Troponin T Type 2 (cardiac) Antibody (BGN/1288/44), Alexa Fluor™ 532, Novus Biologicals™ > Troponin T Type 2 (cardiac) Antibody; Alexa Fluor™ 532; 0.1 mL
Spot an opportunity for improvement?Share a Content Correction