Learn More
USH1G Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA555062
Description
Immunogen sequence: NIWCLDNDYH TPLDMAAMKG HMECVRYLDS IAAKQSSLNP KLVGKLKDKA FREAERRIRE CAKLQRRHHE RMER Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
This gene encodes a protein that contains three ankyrin domains, a class I PDZ-binding motif and a sterile alpha motif. The encoded protein interacts with harmonin, which is associated with Usher syndrome type 1C. This protein plays a role in the development and maintenance of the auditory and visual systems and functions in the cohesion of hair bundles formed by inner ear sensory cells. Mutations in this gene are associated with Usher syndrome type 1G (USH1G).
Specifications
USH1G | |
Polyclonal | |
Unconjugated | |
USH1G | |
ANKS4A; SANS; Scaffold protein containing ankyrin repeats and SAM domain; USH1 protein network component sans; USH1G; Usher syndrome 1G (autosomal recessive); Usher syndrome type-1G protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124590 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q495M9 | |
USH1G | |
Recombinant protein corresponding to Human USH1G | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only