Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water with 0.1% ammonium acetate, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
4 x 2.5 L
4 x 4 L
Packaging:
Glass Bottle
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities

Specifications
Specifications
Packaging | Glass Bottle |
Quantity | 4 x 2.5 L |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Water with 0.1% ammonium acetate, Honeywell Riedel-de Haën™ >
Spot an opportunity for improvement?Share a Content Correction