Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ X-MINI™ Human Selection Kits
Click to view available options
Label Type:
Biotinylated anti-human CD3 Antibody
Biotinylated anti-human CD8 Antibody
Description
- Selected cells are immediately available for downstream applications such as flow cytometry and cell culture expansion.
- Easy to use
- Single-use, column-free and gentle on cells
- High purity: Achieve purities of up to 99%
- Reproducible results (>86%)
- High viability (>97%)
- Process up to 4 x 108 total cells

Specifications
Specifications
Capacity (Metric) | Up to 4 x 108 total cells |
For Use With (Application) | Blood and Blood Products |
Quantity | 1/Pk., 1/Cs. |
Content And Storage | 2°C to 8°C |
Includes | Biotin Mouse Anti-Human Antibody; Corning X-BACS™ Reagent; Corning X-BACS Buffer; Syringe, 10 mL; 21 G x 1.5 in. Needle; Syringe cap |
Label Type | Biotinylated anti-human CD8 Antibody |
Product Type | Cell selection |
Detection Method | Buoyancy Activated Cell Sorting |
Product Line | Corning X-BACS |
Shipping Condition | Room Temperature |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ X-MINI™ Human Selection Kits >
Spot an opportunity for improvement?Share a Content Correction