Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HORIBA Yumizen C1200 Protein Control
Click to view available options
Concentration:
Abnormal concentration for analytes
Normal concentration for analytes
Description
- Used to monitor protein assays
- Human plasma with assigned values determined by calibration with reference material
Specifications
Specifications
Certifications/Compliance | CE Marked |
Content And Storage | 2°C to 8°C |
Concentration | Normal concentration for analytes |
Detectable Analytes | C3, C4, IgA, IgG, IgM, Transferrin (TRF) |
For Use With (Equipment) | Used on Yumizen C1200 Clinical Chemistry Analyzer |
Sample Type | Serum or Plasma |
Type | Control |
Disposable | Single-use |
Quantity | 3 Pk. 1 mL Vials |
Source | Based on Human Plasma |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HORIBA Yumizen C1200 Protein Control >
Spot an opportunity for improvement?Share a Content Correction