Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF239 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310456100UL
Description
ZNF239 Polyclonal specifically detects ZNF239 in Human samples. It is validated for Western Blot.Specifications
ZNF239 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
KIAA1198, zinc finger protein 490 | |
The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF239 (NP_005665). Peptide sequence KGESCSENLQVKLVSDGQELASPLLNGEATCQNGQLKESLDPIDCNCKDI | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
8187 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ZNF239 Rabbit anti-Human, Polyclonal, Novus Biologicals™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction