Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Malate Assay Kit
Click to view available options
Quantity:
1 Kit
Specifications
Specifications
Calibration Range | 1 to 35nm |
Content And Storage | Store the kit at -20°C. |
Description | Malate Assay Kit is used to detecting Malate. |
Detection Method | Colorimetric |
For Use With (Application) | Functional Study |
Includes | 20mL Malate assay buffer, 1 vial Malate enzyme mix (lyophilized), 1 vial WST substrate (lyophilized), 1 vial Malate standard (10μM, lyophilized) |
Quantity | 1 Kit |
Sample Type | Beer, Biological Fluid, Cell Culture Supernatant, Cheese, Food, Fruit, Plasma, Serum, Tissue Culture Supernatant, Urine, Wine |
Type | Detection Assay Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction