Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Ansell AlphaTec™ 2000 Standard Bound Coveralls with Hood, Model 111 Catalog No.: 17999876
Only null left
Add to Cart

Ansell AlphaTec™ 2000 Standard Bound Coveralls with Hood, Model 111

Catalog No. p-7070776
Small
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
This item is not returnable. View return policy

Catalog No. 17-999-876

Supplier: Ansell WH20B9211102

Only null left
Add to Cart
Only null left
Add to Cart
This item is not returnable. View return policy

Coveralls with breathable microporous laminate technology are ideal for a wide range of industrial applications. Previously known as MICROCHEM™ by AlphaTec 2000 Standard.

  • Liquid penetration resistance and barrier to fine particulates
  • Compliant with ASTM F1761 and EN 14126:2003 resistance to penetration of blood, body fluids and blood-borne pathogens
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Low linting reduces the risk of fiber contamination in some critical areas
  • Anti-static properties, tested according to EN1149-5 and AATCC76 (8EMSL test method)
  • Elastic wrists and ankles
  • 2-way front zipper with storm flap
  • Bound seams, finger loops
  • 3-piece hood
  • Applications include pharmaceutical industries, agriculture, cleanrooms, paint spraying, crime scene investigations, veterinary services, food
  • Available in small through 5X-large sizes
  • EN 14126 / ASTM F1671, DIN 32781, EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5B, ISO 17491-4 Type 6B
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 14126 / ASTM F1671, DIN 32781, EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5B, ISO 17491-4 Type 6B

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Small
Gender Unisex
Material Microporous Film Laminate
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Finger Loops
Neck Style Hooded
Hood Type 3-panel, Respirator Fit
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Particle, Liquid, Chemicals, Anti-static
Certifications/Compliance ASTM F1761, AATCC76, ANSI/ISEA 101
Certifications/Compliance (Europe) EN 14126:2003, EN1149-5 , DIN 32781, EN 1073-2, EN ISO 13982-1 Type 5B, ISO 17491-4 Type 6B
Latex Free Yes
Model WH20-B-92-111
For Use With (Application) Life Sciences, Automotive, Metal Fabrication, Food Processing
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell AlphaTec™ 2000 Standard Bound Coveralls with Hood, Model 111 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.