Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BA Antigen (Leu21) Antibody (DF1588) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
Antigen | BA Antigen (Leu21) |
---|---|
Clone | DF1588 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | ||||||
NB254564100
![]() |
Novus Biologicals
NBP254564100UG |
100 μg |
Each for $646.00
|
|
||||||
NB254564200
![]() |
Novus Biologicals
NBP254564200UG |
200 μg | N/A | N/A | N/A | |||||
Description
BA Antigen (Leu21) Monoclonal specifically detects BA Antigen (Leu21) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional, CyTOF-ready.Specifications
BA Antigen (Leu21) | |
1 mg/mL | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
Human leucocytes | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
DF1588 | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with No Preservative | |
IgG1 κ | |
Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BA Antigen (Leu21) Antibody (DF1588) - Azide and BSA Free, Novus Biologicals™