Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCR5 (CD185) Rat anti-Human, BB700, Clone: RF8B2, BD Horizon™
Rat Monoclonal Antibody
$178.00 - $444.00
Specifications
Antigen | CXCR5 (CD185) |
---|---|
Clone | RF8B2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Blue 700 (BB700) |
Description
CXCR5 (CD185) Monoclonal antibody specifically detects CXCR5 (CD185) in Human samples. It is validated for Flow Cytometry.Specifications
CXCR5 (CD185) | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
Rat | |
Human | |
Primary |
RF8B2 | |
Monoclonal | |
Purified | |
RUO | |
IgG2b κ | |
Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CXCR5 (CD185) Rat anti-Human, BB700, Clone: RF8B2, BD Horizon™