Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Specifications
Specifications
Broth Type | Heart Infusion Broth |
Color | Beige |
Form | Powder |
Product Type | Microbiology Media |
For Use With (Application) | Isolation and cultivation of a wide variety of fastidious microorganisms |
Quantity | 500 g |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Bacto™ Heart Infusion Broth >
Spot an opportunity for improvement?Share a Content Correction