Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edge Biologicals Dehydrated Culture Media: Lysine Iron Agar
Click to view available options
:
Lysine Iron Agar; 16 x 125mm; Container: Tube
Description
Laboratory Use Only. Not for use in diagnostic or therapeutic procedures
Specifications
Specifications
Certifications/Compliance | Certificate of analysis |
Content And Storage | 8°C to 12°C |
Form | Dehydrated |
Media Type | Lysine Iron Agar |
Product Type | Agar |
Formulation | Peptone 3g/L, Dextrose 1g/L, Ferric Ammonium Citrate 0.5g/L, Agar 15g/L, Yeast extract 3g/L, Lysine HCl 10g/L, Sodium Thiosulfate 0.04g/L, Bromcresol Purple 0.02g/L, De-ionized Water 1000mL |
For Use With (Application) | Detection Salmonella species |
Packaging Type | Tube |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Edge Biologicals Dehydrated Culture Media: Lysine Iron Agar >
Spot an opportunity for improvement?Share a Content Correction