Learn More
Fisherbrand™ Filter Adapters
(2)
3.8
Average Customer Rating
2 Ratings
4.0
Quality
Does the quality meet your expectations?
3.5
Value
Does the product offer good value for the price?
Average based on verified customer ratings. Learn More
Eliminates boring of special size holes in rubber stoppers
$58.90 - $212.00
Specifications
Autoclavable Autoclavable
Material Neoprene Rubber
For Use With (Equipment) Buchner and fritted Glass funnels, porcelain filters without headpieces
Type Filter Adapter
View More Specs
Products 8
Catalog Number Description Price Quantity & Availability  
Catalog Number Description Price Quantity & Availability  
05-888-107
View Documents
Set of sizes 1-7
Pack of 7 for $58.90
Case of 25 PK for $1,105.00
Only null left
Add to Cart
05-888-101
View Documents
Size 2
Pack of 12 for $131.80
Only null left
Add to Cart
05-888-102
View Documents
Size 3
Pack of 12 for $148.60
Only null left
Add to Cart
05-888-100
View Documents
Size 1
Pack of 12 for $113.50
Only null left
Add to Cart
05-888-104
View Documents
Size 5
Pack of 12 for $156.40
Only null left
Add to Cart
05-888-103
View Documents
Size 4
Pack of 12 for $138.50
Only null left
Edge
Add to Cart
05-888-105
View Documents
Size 6
Pack of 12 for $153.80
Only null left
Edge
Add to Cart
05-888-106
View Documents
Size 7
Pack of 12 for $212.00
Only null left
Add to Cart
Description
Description
Eliminates boring of special size holes in rubber stoppers
Designed to fit Buchner and fritted-glass funnels, porcelain filters without headpieces, and reverse flow cleaning of filter crucibles. Useful in filtration assemblies for supporting funnels, Gooch crucibles, and filter tubes.
  • Neoprene rubber
  • Autoclavable
  • Adapter does not need to be fitted closely around the funnel stem to be functional
  • Can be used singly or nesting with adjacent sizes
  • Available as an adapter set, or in individual sizes
Specifications
Specifications
Autoclavable
Neoprene Rubber
Buchner and fritted Glass funnels, porcelain filters without headpieces
Filter Adapter
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Filter Adapters
Your feedback has been submitted: Thank you for helping us improve our website.