Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fine Chemicals Biosciences Guanine | 73-40-5 | MFCD00071533 | 10g

Supplier:  Sigma Aldrich Fine Chemicals Biosciences G1195010G

Encompass

Guanine | Purity: 98% | Mol Wt: 151.13 | 73-40-5 | MFCD00071533 | 10g

Catalog No. 50-178-8758


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Sigma Aldrich Fine Chemicals Biosciences Guanine | 73-40-5 | MFCD00071533 | 10g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.