Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
MP Biomedicals™ Nigrosin Stain Catalog No.: ICN19459380
Only null left
Add to Cart

MP Biomedicals™ Nigrosin Stain

Catalog No. p-2625553
100 g
Click to view available options
Quantity:
25 g
100 g

Catalog No. ICN19459380

Supplier: MP Biomedicals™ 0219459380

Only null left
Add to Cart
Only null left
Add to Cart

Used as a viable cell staining agent especially in stem cell biology

  • Formula Weight: 737.87
  • CAS # 8005-03-6
  • Synonym: Acid Black 2; C.I. 50420
  • Physical Description: Black crystalline powder
  • Solublility: Soluble in water(1mg/ml-opaque black with purple cast solution)
TRUSTED_SUSTAINABILITY

Specifications

Type Nigrosin
Quantity 100 g
Format Crystal
For Use With (Application) Viable cell staining agent especially in stem cell biology
Storage Requirements 15 to 30°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Nigrosin Stain >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.