Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Chin-Type Gas Masks

Chin-type full face respirators provide protection against particulates, vapors, and gases by way of Ultravue™ facepiece and lightweight, replaceable canister.

Specifications

Color Black
Product Type Gas Mask Respirator
View More Specs

Includes: Mask comes with facepiece and canister.

Products 3
Catalog Number Mfr. No. Cartridge/Filter Price Quantity & Availability  
Catalog Number Mfr. No. Cartridge/Filter Price Quantity & Availability  
19-033-000F
View Documents
MSA™
817064
Ammonia N/A N/A N/A
19-033-000G
View Documents
MSA™
817065
Chlorine N/A N/A N/A
19-033-000H
View Documents
MSA™
817066
Chlorine N/A N/A N/A
Description

Description

  • Face and respiratory protection against particulates, vapors, and gases
  • Ultravue™ facepiece constructed of black Hycar™™ rubber with wraparound poly lens
  • Lightweight, replaceable canister
  • Small, oval-shaped aluminum canister for easy downward head movement
  • Inhalation check valve prevents exhaled air from entering breathing tube
  • Mechanical speaking diaphragm for short-range communication
  • Five adjustable rubber straps and a vinyl neck strap for holding facepiece in standby position
  • Maximum Use Conditions:
    • Entry, 100 times the Permissible Exposure Level (PEL) without exceeding the Immediately Dangerous to Life or Health (IDLH) concentration
    • Escape, contaminant concentrations up to 5000 ppm (0.5% by volume) providing presence of adequate oxygen (19.5% by volume)

Certifications

Canisters are NIOSH approved.

Specifications

Specifications

Black
Gas Mask Respirator
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MSA™ Chin-Type Gas Masks

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.