Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
MilliporeSigma™ Novagen™ Rosetta-gami™ 2 (DE3) Competent Cells Catalog No.: 713514
Only null left
Add to Cart

MilliporeSigma™ Novagen™ Rosetta-gami™ 2 (DE3) Competent Cells

Catalog No. p-7020099
1 mL
Click to view available options
Quantity:
0.4 mL
1 mL

Catalog No. 71-351-4

Supplier: MilliporeSigma™ 713514

Only null left
Add to Cart
Only null left
Add to Cart

Rosetta-Gami 2 host strains allows for enhanced disulfide bond formation and enhanced expression of eukaryotic proteins that contain codons rarely used in E. coli.

Rosetta-gami 2 host strains combine the advantages of Rosetta 2 and Origami 2 strains to alleviate codon bias and enhance disulfide bond formation in the cytoplasm when heterologoous proteins are expressed in E. coli. These trxB/gor mutants are compatible with kanamycin-resistant vectors, and carry the chloramphenicol-resistant pRARE2 plasmid, which supplies seven rare tRNAs. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
TRUSTED_SUSTAINABILITY

Specifications

Efficiency Guaranteed efficiency: >2 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MilliporeSigma™ Novagen™ Rosetta-gami™ 2 (DE3) Competent Cells >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.