Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human Pancreas Membrane Tissue Lysate (Adult Membrane Diabetes)

Click to view available options
Quantity:
0.1 mg
Description
The Human Pancreas Membrane Tissue Lysate (Adult Membrane Diabetes) is derived from human.
Specifications
Specifications
Tissue | Pancreas |
Quantity | 0.1 mg |
Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
Test Specificity | Membrane Protein - Diabetic Disease: Pancreas |
Cellular Fraction | Membrane |
Tissue Condition | Diabetes |
Cross Reactivity | Human |
Host Species | Human |
Product Type | Human Pancreas Membrane Tissue Lysate (Adult Diabetes) |
Content And Storage | -80°C. Avoid freeze-thaw cycles. |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Human Pancreas Membrane Tissue Lysate (Adult Membrane Diabetes) >
Spot an opportunity for improvement?Share a Content Correction