Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Promega NADP/NADPH-Glo™ Assay Catalog No.: PRG9081
Only null left
Add to Cart

Promega NADP/NADPH-Glo™ Assay

Catalog No. p-4762654
10 mL
Click to view available options
Quantity:
10 mL
50 mL

Catalog No. PRG9081

Supplier: Promega G9081

Only null left
Add to Cart
Only null left
Add to Cart

Includes

  • G9081 assay kit components: Reductase (55μL), Reductase Substrate (55μL), NADP Cycling Enzyme (1 vial), NADP Cycling Substrate (275μL), Luciferin Detection Reagent; Reconstitution Buffer (10mL)
  • G9082 assay kit components:Reductase (275μL), Reductase Substrate (275μL), NADP Cycling Enzyme (1 vial), NADP Cycling Substrate (275μL), Luciferin Detection Reagent; Reconstitution Buffer (50mL)

The NAD/NADH-Glo™ Assay is a bioluminescent, homogeneous single-reagent-addition assay for detecting total oxidized and reduced nicotinamide adenine dinucleotides and determining their ratio in biological samples or in defined enzyme reactions.

  • Bioluminescent Assays to Detect NAD(P)+ and NAD(P)H Levels in Cells
  • Homogeneous, single-reagent-addition assays
  • Easily adapted for high throughput screening formats

Specifications

Quantity 10 mL
Product Type NADP/NADPH-Glo assay kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Promega NADP/NADPH-Glo™ Assay >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.