Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Thermo Scientific™ Simmons Citrate Agar Catalog No.: R060494
Only null left
Add to Cart

Thermo Scientific™ Simmons Citrate Agar

Catalog No. p-4524006
100 x 5 mL
Click to view available options
Quantity:
20 x 5 mL
100 x 5 mL
This item is not returnable. View return policy

Catalog No. R060494

Supplier: Thermo Scientific™ R060494

Only null left
Add to Cart
Only null left
Add to Cart
This item is not returnable. View return policy

Differentiate gram-negative bacteria based on citrate utilization using Simmons Citrate Agar.

Differentiate gram-negative bacteria based on citrate utilization using Thermo Scientific™ Remel™ Simmons Citrate Agar. In 1923, Koser developed a liquid medium to differentiate Escherichia coli and Enterobacter aerogenes1. The medium contained an inorganic ammonium salt and sodium citrate that provided a single nitrogen source and a single carbon source respectively. In 1926, this formulation was modified by Simmons who added 1.5% agar and brom thymol blue indicator2.

Gram-negative bacteria that can utilize ammonium dihydrogen phosphate as a sole nitrogen source and sodium citrate as a sole carbon source will grow on Citrate Agar (Simmons). These organisms extract nitrogen from the ammonium salt by breaking it down to form ammonia, leading to alkalization of the medium. This causes the brom thymol blue indicator to change color from green to blue.

  • Ready to use – Convenience of prepared medium

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, USDA.
Description Simmons Citrate Agar
Format 15mm x 103mm Tube - Slant
Quantity 100 x 5 mL

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number246711 Certificate TypeCertificate of Analysis Date03/20/2025 Catalog NumberR060494
Lot Number218191 Certificate TypeCertificate of Analysis Date01/11/2025 Catalog NumberR060494
Lot Number187408 Certificate TypeCertificate of Analysis Date11/04/2024 Catalog NumberR060494
Lot Number150123 Certificate TypeCertificate of Analysis Date07/25/2024 Catalog NumberR060494
Lot Number112168 Certificate TypeCertificate of Analysis Date05/09/2024 Catalog NumberR060494
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Simmons Citrate Agar >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.