Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
MP Biomedicals™ SPINeasy™ Plasmid Midiprep Kit Catalog No.: MP116539025
Only null left
Add to Cart

MP Biomedicals™ SPINeasy™ Plasmid Midiprep Kit

Catalog No. MP116539025
25 Preps
Click to view available options
Quantity:
25 Preps

Catalog No. MP116539025

Supplier: MP Biomedicals™ 116539025

Only null left
Add to Cart
Only null left
Add to Cart

Spin column technology for fast and efficient purification of both high and low copy plasmids from bacterial cultures. High-quality plasmid DNA is free of gDNA contamination following a modified alkaline lysis method.

  • SPINeasy Plasmid Midiprep Kit is a silica membrane spin column kit for the purification of high-copy and low-copy number plasmid DNA from 25 to 50 mL of transformed bacterial cultures.
  • Using our specially formulated buffers, bacterial cells are disrupted by alkaline lysis to release plasmid DNA.
  • Cell debris, such as chromosomal DNA and proteins, are precipitated by neutralization and cleared through centrifugation. Subsequently, plasmid DNA in the supernatant is adsorbed on a silica membrane, washed, and eluted in a convenient process using spin column technology.
  • Up to 1 mg of plasmid DNA can be purified through this simple workflow, which is immediately ready for routine molecular biology applications, including cloning, in-vitro transcription, PCR, and restriction digestion.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Isolation Technology Spin Column
Sample Type 25 to 50 mL Bacterial (E. coli) Cultures
Includes 60 mL Resuspension Buffer SD1, 60 mL Alkaline Lysis Buffer SD2, 90 mL Neutralization Buffer SD3, 70 mL Wash Buffer DW1, 12 mL Wash Buffer DW2, 30 mL Elution Buffer SD, 600 µL RNase A, 25 x Column SD with collection tubes
Quantity 25 Preps

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ SPINeasy™ Plasmid Midiprep Kit >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.