Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ TFC 007

Potent hematopoietic prostaglandin D synthase (H-PGDS) inhibitor
$209.00 - $870.50
Specifications
Purity | >98% |
---|---|
Product Type | TFC 007 |
Molecular Weight (g/mol) | 487.55 |
Inhibitors | TFC 007 |
Description
Potent hematopoietic prostaglandin D synthase (H-PGDS) inhibitor (IC50 = 83 nM). Inhibits PGD2 production and late phase nasal blockage in an animal model of allergic rhinitis. Exhibits negligible effects on COX-1, COX-2, 5-LOX, LTC4 synthase, TXA2 synthase and microsomal prostaglandin E2 synthase.Specifications
>98% | |
TFC 007 | |
487.55 | |
TFC 007 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ TFC 007