Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Oatmeal Agar
Click to view available options
:
Oatmeal Agar; 500g
Specifications
Specifications
Certifications/Compliance | CE |
Content And Storage | Refrigerate |
Form | Dehydrated |
Media Type | Oatmeal Agar |
Product Type | Agar |
Formulation | Oatmeal 60g/L, Agar 12.5g/L |
For Use With (Application) | Cultivation of fungi, particularly for macrospore formation |
Packaging Type | Bottle |
Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction