Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conductivity Standard, 1000 μS/cm at 25°C (495 ppm TDS as NaCl), Ricca Chemical
SDP

Catalog No. p-2915804
Click to view available options
Quantity:
100 mL
500 mL
1 L
20 x 20 mL
4 L
Packaging:
Natural Poly Bottle
Pack

Specifications

Chemical Name or Material Conductivity/TDS Standard
Packaging Pack
Name Note 1mS/cm (1000μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 98.0, 0.05
CAS Max % 100.0, 0.05
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 495ppm as NaCl
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.