Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Meloxicam Catalog No.: AAJ6063506
Only null left
Add to Cart

Meloxicam

Catalog No. AAJ6063506
5 g
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAJ6063506

Supplier: Thermo Scientific Chemicals J6063506

Only null left
Add to Cart
Only null left
Add to Cart
Request Bulk or Custom Formats

CAS: 71125-38-7 | C14H13N3O4S2 | 351.40 g/mol

Meloxicam is developed for the treatment of rheumatoid arthritis and osteoarthritis. It is currently for the treatment of osteoarthritis and rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (22 mg/ml), DMSO (25 mg/ml), DMF, acetone(slightly soluble), ethanol(slightly soluble), and methanol(slightly soluble).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71125-38-7
Molecular Formula C14H13N3O4S2
Molecular Weight (g/mol) 351.40
MDL Number MFCD00868752
InChI Key DWMREKMVXIFPFM-ACCUITESSA-N
Synonym meloxicam, mobic, metacam, movalis, meloxicamum, mobicox, mobec, meloxicam mobic, movatec, parocin
PubChem CID 54677470
ChEBI CHEBI:6741
IUPAC Name (3E)-3-{hydroxy[(5-methyl-1,3-thiazol-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λ⁶,2-benzothiazine-1,1,4-trione
SMILES CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O

Specifications

Melting Point 255°C
Color Yellow
Quantity 5 g
UN Number UN2811
Merck Index 14,5826
Solubility Information Soluble in water (22mg/ml),DMSO (25mg/ml),DMF,acetone(slightly soluble),ethanol(slightly soluble),and methanol(slightly soluble).
Formula Weight 351.39
Physical Form Powder
Chemical Name or Material Meloxicam

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS04L013 Certificate TypeCertificate of Analysis Date06/05/2025 Catalog NumberJ6063514,J6063506
Lot NumberU27K050 Certificate TypeCertificate of Analysis Date10/03/2024 Catalog NumberJ6063514,J6063506
Lot NumberQ30K006 Certificate TypeCertificate of Analysis Date05/07/2024 Catalog NumberJ6063514,J6063506
Lot NumberQ05K045 Certificate TypeCertificate of Analysis Date04/17/2024 Catalog NumberJ6063514,J6063506
Lot NumberU02I029 Certificate TypeCertificate of Analysis Date08/11/2022 Catalog NumberJ6063514,J6063506
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber DL0702000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Meloxicam >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.