Learn More
Medchemexpress LLC HY-129217 500mg Medchemexpress, Naringinase CAS:9068-31-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY129217500MG
Encompass_Preferred
Medchemexpress, HY-129217 500mg Naringinase CAS:9068-31-9 Naringinase, a hydrolytic enzymatic complex, possesses the activity of both α-L-rhamnosidase and β-D-glucosidase. Naringinase has wide occurrence in nature. Naringinase can be used in the biotransformation of steroids, antibiotics, and mainly on glycosides hydrolysis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1200
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Medchemexpress LLC HY-129217 500mg Medchemexpress, Naringinase CAS:9068-31-9 Purity:>98%
Your feedback has been submitted: Thank you for helping us improve our website.