Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Noble Agar, Ultrapure Catalog No.: AAJ1090722
Only null left
Add to Cart

Noble Agar, Ultrapure

Catalog No. p-7076792
100 g
Click to view available options
Quantity:
100 g
500 g
1 kg

Catalog No. AAJ1090722

Supplier: Thermo Scientific J1090722

Only null left
Add to Cart
Only null left
Add to Cart
Request Bulk or Custom Formats

Gelling agent essentially free of impurities. Thermo Scientific™ Noble Agar (Ultrapure) is mainly used in biochemistry and molecular biology procedures where increased purity is required.

Gelling agent essentially free of impurities. Used mainly in biochemistry and molecular biology procedures wherever increased purity is required.

Typical Agar Preparation:

  • Plates: 15 g/L of media; 30-35 ml per 90 mm plate
  • Top Agar: 7 g/L of media

Autoclave for 20 minutes and swirl gently to evenly distribute the agar. If antibiotics are to be added, allow the agar to cool to 50°C. When pouring plates, gently swirl the medium. A Bunsen burner flame can be passed over the surface of the plates to remove any air bubbles. After the medium has hardened, invert the plates and store at 4°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Format: Pale cream powder
  • CAS: 9002-18-0
  • MDL Number: MFCD00081288

Typical Agar Preparation
Plates: 15g/L of media; 30 to 35mL per 90mm plate

Top Agar: 7g/L of media

Autoclave for 20 minutes and swirl gently to evenly distribute the agar. If antibiotics are to be added, allow the agar to cool to 50°C. When pouring plates, gently swirl the medium. A Bunsen burner flame can be passed over the surface of the plates to remove any air bubbles. After the medium has hardened, invert the plates and store at 4°C

TRUSTED_SUSTAINABILITY

Specifications

Color Pale cream
Form Powder
Grade Ultrapure
Product Type Agar
For Use With (Application) Used as a Gelling Agent in Biochemistry and Molecular Biology Procedures Where Increased Purity is Required.
Packaging Type Plastic Bottle
Quantity 100 g

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number191928 Certificate TypeCertificate of Analysis Date06/13/2025 Catalog Number
Lot Number213191 Certificate TypeCertificate of Analysis Date06/13/2025 Catalog Number
Lot Number235322 Certificate TypeCertificate of Analysis Date06/13/2025 Catalog Number
Lot Number218252 Certificate TypeCertificate of Analysis Date06/13/2025 Catalog Number
Lot Number244878 Certificate TypeCertificate of Analysis Date06/13/2025 Catalog Number
EINECSNumber 232-658-1
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Noble Agar, Ultrapure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.