Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Calibrators: Total B-HCG II

Click to view available options
Quantity:
3 Sets/Pk.
Specifications
Specifications
Content And Storage | 2°C to 8°C |
For Use With (Equipment) | VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system |
Sample Type | Human Serum and Plasma (Heparin and EDTA) |
Type | Total B-HCG II Calibrator |
Includes | VITROS™ Total beta-hCG II calibrators 1, 2 and 3, (freeze-dried, recombinant HCG in Human Plasma with antimicrobial agent, reconstitution volume 1mL), lot calibration card, protocol card, 24 calibrator bar code labels (8 for each calibra |
Intended Use | Quantitative measurement of Human Chorionic Gonadotropin (hCG) and its beta-subunit |
Level | 3 |
Lyophilized | Yes |
Quantity | 3 Sets/Pk. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ortho-Clinical Diagnostics™ VITROS™ Calibrators: Total B-HCG II >
Spot an opportunity for improvement?Share a Content Correction