Learn More
Medchemexpress LLC HY-136532 5mg Medchemexpress, ZT-1a CAS:212135-62-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1365325MG
Encompass_Preferred
Medchemexpress, HY-136532 5mg ZT-1a CAS:212135-62-1 ZT-1a is a potent, non-ATP-competitive and selective SPAK inhibitor. ZT-1a inhibits SPAK activity with IC50s of 44.3, 35.0, 46.7 μM at ATP concentrations of 0.01, 0.1 and 1 mM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0450
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Medchemexpress LLC HY-136532 5mg Medchemexpress, ZT-1a CAS:212135-62-1 Purity:>98%
Your feedback has been submitted: Thank you for helping us improve our website.