Learn More
HORIBA Pentra™ XL 80 Hematology System
Utilizes flow cytometry using principles of impedance, cytochemistry and measurement of light absorbance (DHSS); Multi Distribution Sampling System (MDSS) microsampling
Supplier:  HORIBA 5300800011
Catalog No. 23-500-106
Request a Quote
Item Discontinued This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
Description
Description
Pentra XL 80 System Features
  • Complete blood-cell count and differential leukocyte count with 20 parameters
  • 80 samples/hour
  • 5-part differential
  • Automatic sample handling
  • Touch-screen monitor and interactive software
Pentra XL 80 Enhanced Features
  • Customized Dilution Ratio
  • Delta Check flagging
Specifications
Specifications
ABX Pentra™ XL 80
30 μL (CBC), 53 μL (CBC + DIFF)
Printer
22.4 in.
21.5 in.
32.3 in.
100/240V 50/60Hz
55 kg
Utilizes flow cytometry using principles of impedance, cytochemistry and measurement of light absorbance (DHSS) multi distribution sampling system (MDSS) microsampling
80 Samples/Hour
1 Year
57 cm
54 cm
82 cm
122 lb.
20 Parameters, WBC, NEU# and NEU%, LYM# and LYM%, MON# and MON%, EOS# and EOS%, BAS# and BAS%, RBC, HGB, HCT, MCV, MCH, MCHC, RDW, PLT, MPV
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HORIBA Pentra™ XL 80 Hematology System > Pentra XL 80
Your feedback has been submitted: Thank you for helping us improve our website.